Tetrahydrofuran 99+% stabilisiert, reinst

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Butylene oxide, THF, Diethylene oxide

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

176630010 176630025 176630250 176630050 176630100
ACRO176630010EA 66.9 CHF
ACRO176630010 ACRO176630025 ACRO176630250 ACRO176630050 ACRO176630100
Tetrahydrofuran 99+% stabilisiert, reinst
Tetrahydrofuran
Formel: C₄H₈O
Siedepunkt: 66 °C (760 mmHg)
Melting Pt: –108.4 °C
Dichte: 0.88 g/cm³
Flash-Pt: –21 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00005356
CAS-Nummer: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance Clear liquid
Water ≤0.05 % (Coulometric)
GC ≥99.0 %
Color scale ≤10 APHA
Stabilizer 200 to 400 ppm BHT
Residue after evaporation ≤0.005 %
Peroxides ≤0.0025 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR