4-tert-Butylbenzoylchlorid 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: 4-tert-Butylbenzoylchloride

169100050 169100250 169101000
ACRO169100050EA 27.6 CHF
ACRO169100050 ACRO169100250 ACRO169101000
4-tert-Butylbenzoylchlorid 98%
4-tert-Butylbenzoylchlorid
Formel: C₁₁H₁₃ClO
MW: 196.68 g/mol
Siedepunkt: 135 °C (20 mmHg)
Dichte: 1 g/cm³
Flash-Pt: 87 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000695
CAS-Nummer: 1710-98-1
EINECS: 216-973-1
UN: 2922

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5354 to 1.5374 (20°C, 589 nm)
Titration RCOCl ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR