L-Methionin 98+%

Lieferant: Thermo Fisher Scientific

Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid

166160250 166161000 166160025 166165000
ACRO166160250EA 36.7 CHF
ACRO166160250 ACRO166161000 ACRO166160025 ACRO166165000
L-Methionin 98+%
L-Methionin
Formel: C₅H₁₁NO₂S
Siedepunkt: 278 °C (1013 hPa)
Melting Pt: 276…279 °C (dec.)
Dichte: 1,373 g/cm³ (20 °C)
Lager Temperatur: Kühlschrank
MDL Number: MFCD00063097
CAS-Nummer: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004

Order Now

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Sulfated ash ≤0.4 %
TLC ≤0.5 % of any individual impurity
pH 5.5 to 6.5 (1 % in water)
Specific optical rotation +22.4° to +24.7° (20°C, 589 nm) (c=2, 6 N HCl)
Titration with HClO4 98.0 to 102.0 % (on dried basis)
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤300 ppm
Iron (Fe) ≤30 ppm
Heavy metals (as Pb) ≤15 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR