Propionamid 97%

Lieferant: Thermo Fisher Scientific
Achtung

162671000 162670250
ACRO162671000EA 110 CHF
ACRO162671000 ACRO162670250
Propionamid 97%
Propionamid
Formel: C₃H₇NO
Siedepunkt: 213 °C (760 mmHg)
Melting Pt: 79…83 °C
Dichte: 1,1 g/cm³ (20 °C)
Flash-Pt: >100 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008039
CAS-Nummer: 79-05-0
EINECS: 201-172-1
Merck Index: 12,08009

Order Now

Spezifikation Testergebnisse

Appearance Colorless to white to light yellow Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
Melting point 77 to 83 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR