Benzamid 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Benzoic acid amide

159331000 159335000 159330025
ACRO159331000EA 31 CHF
ACRO159331000 ACRO159335000 ACRO159330025
Benzamid 99%
Benzamid
Formel: C₇H₇NO
MW: 121.14 g/mol
Siedepunkt: 228 °C (760 mmHg)
Melting Pt: 125…129 °C
Flash-Pt: 180 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007968
CAS-Nummer: 55-21-0
EINECS: 200-227-7
Merck Index: 13,01059

Order Now

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 125 to 129 °C
HPLC ≥98.5 %
UV at 280 nm A: ≤0.05
Titration with HClO4 99.4 to 100.6 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤0.0005 %
Sodium (Na) ≤0.02 %
Heavy metals (as Pb) ≤5 ppm

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR