Diisobutylamin 99%

Lieferant: Thermo Fisher Scientific
Gefahr

159070010 159075000 159070025
ACRO159070010EA 61 CHF
ACRO159070010 ACRO159075000 ACRO159070025
Diisobutylamin 99%
Diisobutylamin
Formel: C₈H₁₉N
MW: 129.25 g/mol
Siedepunkt: 137…139 °C (760 mmHg)
Melting Pt: –77 °C
Dichte: 0.74 g/cm³
Flash-Pt: 26 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008930
CAS-Nummer: 110-96-3
EINECS: 203-819-3
UN: 2361
ADR: 3 III

Order Now

Spezifikation Testergebnisse

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4071 to 1.4091 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %
Color scale ≤15 APHA
Sulfate (SO4) ≤0.1 %
Arsenic (As) ≤3 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤2 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR