n-Butylisocyanid 98+%

Lieferant: Thermo Fisher Scientific
Gefahr

158380010 158380050
ACRO158380010EA 82.6 CHF
ACRO158380010 ACRO158380050
n-Butylisocyanid 98+%
n-Butylisocyanid
Formel: C₅H₉N
Dichte: 0.79 g/cm³
Lager Temperatur: Kühlschrank
MDL Number: MFCD00000008
CAS-Nummer: 2769-64-4
EINECS: 220-458-7
UN: 1935
ADR: 6.1,II

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3950 to 1.3970 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR