Anilin 99.8%, rein

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Aminobenzene, Phenylamine

158190010 158190050 158190025 158191000
ACRO158190010EA 57 CHF
ACRO158190010 ACRO158190050 ACRO158190025 ACRO158191000
Anilin 99.8%, rein
Anilin
Formel: C₆H₅NH₂
Siedepunkt: 181…185 °C (760 mmHg)
Melting Pt: –6.2 °C
Dichte: 1.0213 g/cm³
Flash-Pt: 76 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007629
CAS-Nummer: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1,II
Merck Index: 13,00661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc5b5071f5a578057c8a7c1c4855ca964c4d840&vid=4f9c118a47c84e5b067d971f00a4eee6dc846d80&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless to yellow to orange to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5845 to 1.5875 (20°C, 589 nm)
GC ≥99.75 %
Sulfate (SO4) ≤0.03 %
Calcium (Ca) ≤0.005 %
Sodium (Na) ≤0.02 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR