EDTA Trinatriumsalz Hydrat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Trisodium hydrogen ethylenediaminetetraacetate hydrate, Ethylenediaminetetra-acetic acid trisodium salt hydrate, Edetic acid trisodium salt hydrate, Ethylenedinitrilotetra-acetic acid trisodium salt hydrate

147865000
ACRO147865000EA 101 CHF
ACRO147865000
EDTA Trinatriumsalz Hydrat 98%
EDTA Trinatriumsalz Hydrat
Formel: C₁₀H₁₃N₂Na₃O₈·H₂O
MW: 376.21 g/mol
Melting Pt: 300 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149675
CAS-Nummer: 85715-60-2
EINECS: 205-758-8

Order Now

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Complexometric ≥97.5 % (corrected for water)
Water ≤10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR