Triphenylamin 99+%

Lieferant: Thermo Fisher Scientific
Achtung

140310250 140311000
ACRO140310250EA 76.5 CHF
ACRO140310250 ACRO140311000
Triphenylamin 99+%
Triphenylamin
Formel: C₁₈H₁₅N
MW: 245.32 g/mol
Siedepunkt: 347…348 °C (760 mmHg)
Melting Pt: 125…127 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003020
CAS-Nummer: 603-34-9
EINECS: 210-035-5

Order Now

Spezifikation Testergebnisse

Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 125 to 127 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR