n-Tridecan 99+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Tridecane

139510250 139511000 139515000
ACRO139510250EA 51.3 CHF
ACRO139510250 ACRO139511000 ACRO139515000
n-Tridecan 99+%
n-Tridecan
Formel: H₃C(CH₂)₁₁CH₃
Siedepunkt: 234 °C (760 mmHg)
Melting Pt: 5…4 °C
Dichte: 0.75 g/cm³
Flash-Pt: 79 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008979
CAS-Nummer: 629-50-5
EINECS: 211-093-4
ADR: 3,III

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4260 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR