Isonicotinaldehyd 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Pyridine-4-carboxaldehyde, 4-Pyridinecarboxaldehyde, 4-Pyridinecarbaldehyde

131840250 131845000 131841000
ACRO131840250EA 48.7 CHF
ACRO131840250 ACRO131845000 ACRO131841000
Isonicotinaldehyd 98%
Isonicotinaldehyd
Formel: C₆H₅NO
Siedepunkt: 77…78 °C (12 mmHg)
Dichte: 1.138 g/cm³
Flash-Pt: 82 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00006425
CAS-Nummer: 872-85-5
EINECS: 212-832-3
UN: 1760
ADR: 8,III

Order Now

Spezifikation Testergebnisse

Appearance Clear yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5420 to 1.5450 (20°C, 589 nm)
GC ≥97.5 %
Chloride (Cl) ≤0.003 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR