2-Pyrazincarbonsäure 99%

Lieferant: Thermo Fisher Scientific

Synonyms: Pyrazinecarboxylic acid, 2-Pyrazinecarboxylic Acid

131710250 131711000
ACRO131710250EA 34.8 CHF
ACRO131710250 ACRO131711000
2-Pyrazincarbonsäure 99%
Pyrazine-2-carboxylic acid
Formel: C₅H₄N₂O₂
MW: 124.1 g/mol
Siedepunkt: 313
Melting Pt: 222…225 °C
MDL Number: MFCD00006130
CAS-Nummer: 98-97-5
EINECS: 202-718-1
Merck Index: 12,08143

Order Now

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 222 to 225 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR