1-Naphthylisothiocyanat 98%

Lieferant: Thermo Fisher Scientific
Gefahr

128240050 128240100
ACRO128240050EA 94.6 CHF
ACRO128240050 ACRO128240100
1-Naphthylisothiocyanat 98%
1-Naphthylisothiocyanat
Formel: C₁₁H₇NS
Siedepunkt: 100 °C (0.2 mmHg)
Melting Pt: 55.5…57 °C
Dichte: 1,81 g/cm³ (20 °C)
MDL Number: MFCD00003882
CAS-Nummer: 551-06-4
EINECS: 208-990-8
UN: 2811
ADR: 6.1,III
Merck Index: 12,06498

Order Now

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 57 °C
GC ≥97.5 %
Sulfate (SO4) ≤0.002 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 % (5 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR