2-Pentanon 99%, pure

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Methyl Propyl Ketone

123502500 123500025 123500010 123500100
ACRO123502500EA 34 CHF
ACRO123502500 ACRO123500025 ACRO123500010 ACRO123500100
2-Pentanon 99%, pure
2-Pentanon
Formel: C₅H₁₀O
MW: 86.13 g/mol
Siedepunkt: 100…103 °C (760 mmHg)
Melting Pt: –78 °C
Dichte: 0.809 g/cm³
Flash-Pt: 7 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009400
CAS-Nummer: 107-87-9
EINECS: 203-528-1
UN: 1249
ADR: 3 II
Merck Index: 12,06193

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3890 to 1.3910 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR