2-Imidazolidinon Hemihydrat 99+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 2-Imidazolidone hemihydrate

122040025 122040250 122045000 122040100
ACRO122040025EA 184 CHF
ACRO122040025 ACRO122040250 ACRO122045000 ACRO122040100
2-Imidazolidinon Hemihydrat 99+%
2-Imidazolidinon Hemihydrat
Formel: C₃H₆N₂O·0.5H₂O
Siedepunkt: >100 °C (760 mmHg)
Melting Pt: 58 °C
Flash-Pt: 265 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD02662355
CAS-Nummer: 121325-67-5
EINECS: 204-436-4

Order Now

Spezifikation Testergebnisse

Appearance White to light yellow Adhering crystalline powder or crystals
Infrared spectrum Conforms
Water ≤13 %
GC ≥99.0 %
Elemental Analysis 5.1 to 5.6 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR