Glycin (H-Gly-OH) 98%, extra_pure

Lieferant: Thermo Fisher Scientific

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

120070010 120070050 120072500 120070250
ACRO120070010EA 116 CHF
ACRO120070010 ACRO120070050 ACRO120072500 ACRO120070250
Glycin (H-Gly-OH) 98%, extra_pure
Glycin (H-Gly-OH)
Formel: NH₂CH₂COOH
MW: 75.07 g/mol
Siedepunkt: 181
Melting Pt: 240 °C
Dichte: 1.595 g/cm³
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008131
CAS-Nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
pH 5.5 to 6.5 (5 % in water)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR