Ferrocen 98%

Lieferant: Thermo Fisher Scientific
Achtung

119141000 119145000 119140025
ACRO119141000EA 72.8 CHF
ACRO119141000 ACRO119145000 ACRO119140025
Ferrocen 98%
Ferrocen
Formel: C₁₀H₁₀Fe
MW: 186.04 g/mol
Siedepunkt: 249 °C (760 mmHg)
Melting Pt: 173…176 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00001427
CAS-Nummer: 102-54-5
EINECS: 203-039-3
UN: 1325
ADR: 4.1 II
Merck Index: 13,04070

Order Now

Spezifikation Testergebnisse

Appearance Orange-brown Crystalline powder or crystals (May become lumpy on storage)
Infrared spectrum Conforms
Melting point 173 to 176 °C
GC ≥97.5 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR