Dimethylphthalat 99%

Lieferant: Thermo Fisher Scientific

Synonyms: Phthalic acid dimethyl ester

116640010 116642500 116640025 116640250
ACRO116640010EA 64.5 CHF
ACRO116640010 ACRO116642500 ACRO116640025 ACRO116640250
Dimethylphthalat 99%
Dimethylphthalat
Formel: C₁₀H₁₀O₄
Siedepunkt: 282 °C (760 mmHg)
Melting Pt: 6 °C
Dichte: 1.19 g/cm³
Flash-Pt: 150 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008425
CAS-Nummer: 131-11-3
EINECS: 205-011-6
Merck Index: 13,03281

Order Now

Spezifikation Testergebnisse

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5140 to 1.5160 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤20 APHA
Iron (Fe) ≤0.002 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.02 %
Strontium (Sr) ≤0.7 %
Heavy metals (as Pb) ≤10 ppm
Water soluble titr. base ≤0.002 meq/g
Insoluble matter ≤0.015 % (in dilute HCl)
Oxidizing substance (as NO3) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR