Dimercaprol 97,0-100,5%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Dimercapto-2,3 propanol-1, Dithioglycerine, 2,3-Dimercaptopropanol, BAL, Dimercaprol, 2,3-Dimercaptopropan-1-ol, 2,3-Dimercaptopropane-1-ol

115300050 115300250
ACRO115300050EA 162 CHF
ACRO115300050 ACRO115300250
Dimercaprol 97,0-100,5%
2,3-Dimercapto-1-propanol
Formel: C₃H₈OS₂
Siedepunkt: 120 °C (15 mmHg)
Melting Pt: 77 °C
Dichte: 1.243 g/cm³
Flash-Pt: >112 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00004864
CAS-Nummer: 59-52-9
EINECS: 200-433-7
UN: 2810
ADR: 6.1,III
Merck Index: 13,03234
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=745c14e3244d75cab02b536ae9d18ec21829aeba&vid=345ebd39845f4c372f03844bee87b4d8c249ec2b&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5700 to 1.5750 (20°C, 589 nm)
Titration Iodimetric 97.0 to 100.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR