1,3-Diamino-2-propanol 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 1,3-Diamino-2-hydroxypropane, 1,3-Diaminopropan-2-ol

112220050 112220250
ACRO112220050EA 60.4 CHF
ACRO112220050 ACRO112220250
1,3-Diamino-2-propanol 95%
1,3-Diamino-2-propanol
Formel: C₃H₁₀N₂O
MW: 90.13 g/mol
Siedepunkt: 114 °C (5 mmHg)
Melting Pt: 39…45 °C
Flash-Pt: >110 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00008142
CAS-Nummer: 616-29-5
EINECS: 210-474-2

Order Now

Spezifikation Testergebnisse

Appearance White to yellow Low melting solid
Infrared spectrum Conforms
Titration with HClO4 ≥94 %
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR