Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride 99+%

Lieferant: Thermo Fisher Scientific

Synonyms: Bz-Arg-OEt.HCl, Nα-Benzoyl-L-arginine Ethyl Ester HCl

105700025 105700100 105700500
ACRO105700025EA 81.4 CHF
ACRO105700025 ACRO105700100 ACRO105700500
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride 99+%
Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride

CAS No.: 2645-08-1

Formel: C₁₅H₂₂N₄O₃
MW: 342.82 g/mol
Melting Pt: 129…136 °C
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00012579
CAS-Nummer: 2645-08-1

Order Now

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless
TLC ≥99.0 %
Specific optical rotation -15.0° to -19.0° (24°C, 589 nm) (c=2, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR