Anti-Theophylline Mouse Monoclonal Antibody [clone: 12-127.1]

Lieferant: Abnova
MAB2232
ABNOMAB2232EA 483 CHF
ABNOMAB2232
Anti-Theophylline Mouse Monoclonal Antibody [clone: 12-127.1]
Antikörper
Mouse monoclonal antibody raised against Theophylline.

Type: Primary
Antigen: Theophylline
Clonality: Monoclonal
Clone: 12-127.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR