Anti-CTSL Mouse Monoclonal Antibody [clone: 2H7]

Lieferant: Abnova
MAB2047
ABNOMAB2047EA 483 CHF
ABNOMAB2047
Anti-CTSL Mouse Monoclonal Antibody [clone: 2H7]
Antikörper
Mouse monoclonal antibody raised against partial recombinant CTSL1.

Type: Primary
Antigen: CTSL
Clonality: Monoclonal
Clone: 2H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR