Anti-LAT Mouse Polyclonal Antibody

Lieferant: Abnova
H00027040-A01
ABNOH00027040-A01EA 312 CHF
ABNOH00027040-A01
Anti-LAT Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length recombinant LAT.

Recommended Dilutons: Western Blot (1:500 to 1:1000).

Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR