Anti-UBE2C Mouse Monoclonal Antibody [clone: 1D8]

Lieferant: Abnova
H00011065-M12
ABNOH00011065-M12EA 477 CHF
ABNOH00011065-M12
Anti-UBE2C Mouse Monoclonal Antibody [clone: 1D8]
Antikörper
Type: Primary
Antigen: UBE2C
Clonality: Monoclonal
Clone: 1D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR