Anti-MAPK1IP1L Rabbit Polyclonal Antibody

Lieferant: Abgent
AP9730C
ABGEAP9730CEA 412 CHF
ABGEAP9730C
Anti-MAPK1IP1L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAPK1IP1L (mitogen-activated protein kinase 1 interacting protein 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR