Anti-EXT2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP9401C
ABGEAP9401CEA 412 CHF
ABGEAP9401C
Anti-EXT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EXT2 (exostosin glycosyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR