Anti-OGG1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP6931B
ABGEAP6931BEA 412 CHF
ABGEAP6931B
Anti-OGG1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OGG1 (8-oxoguanine DNA glycosylase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR