Anti-MMP13 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP6197A
ABGEAP6197AEA 412 CHF
ABGEAP6197A
Anti-MMP13 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MMP13 (matrix metallopeptidase 13 (collagenase 3))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

TECHNISCHE DATEN

Antigen-Symbol MMP13
Environmentally Preferable
Antigen-Name Matrix Metalloproteinase 13
Antigen-Synonyme MMP-13|CLG3|Collagenase 3|matrix metallopeptidase 13|MANDP1
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Isotyp
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Durchflusszytometrie Yes
Epitop C-Terminal (335–349 aa)
Kreuzadsorption No
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR