Anti-MFAP1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP1928B
ABGEAP1928BEA 412 CHF
ABGEAP1928B
Anti-MFAP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MFAP1 (microfibrillar-associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

TECHNISCHE DATEN

Antigen-Symbol MFAP1
Antigen-Name Microfibrillar-Associated Protein 1
Environmentally Preferable
Antigen-Synonyme RB1|pRb|OSRC|pp110|PP110|P105-RB|RB|Retinoblastoma-associated protein|p105-Rb
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Isotyp
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Epitop N-terminal
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR