Anti-ARMC1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP17956B
ABGEAP17956BEA 412 CHF
ABGEAP17956B
Anti-ARMC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARMC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR