Anti-FAU Rabbit Polyclonal Antibody

Lieferant: Abgent
AP1600B
ABGEAP1600BEA 412 CHF
ABGEAP1600B
Anti-FAU Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FAU (Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

TECHNISCHE DATEN

Antigen-Symbol FAU
Antigen-Name Chromosome 13 open reading frame 39
Environmentally Preferable
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Isotyp
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Epitop C-terminal
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR