Anti-HIST1H2AA Rabbit Polyclonal Antibody

Lieferant: Abgent
AP12399A
ABGEAP12399AEA 412 CHF
ABGEAP12399A
Anti-HIST1H2AA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIST1H2AA (histone cluster 1, H2aa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR