Anti-NBPF5P Rabbit Polyclonal Antibody

Lieferant: Abgent
AP12364B
ABGEAP12364BEA 412 CHF
ABGEAP12364B
Anti-NBPF5P Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NBPF5P (neuroblastoma breakpoint family, member 5, pseudogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

TECHNISCHE DATEN

Antigen-Symbol NBPF5
Environmentally Preferable
Antigen-Name Neuroblastoma Breakpoint Family member 5
Antigen-Synonyme p18-INK4C|INK4C|p18
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Isotyp
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Epitop C-terminal
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR