Anti-KLF4 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP11958B
ABGEAP11958BEA 412 CHF
ABGEAP11958B
Anti-KLF4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLF4 (Kruppel-like factor 4 (gut))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR