Anti-PAX6 Mouse Monoclonal Antibody

Lieferant: Abgent
AM1871A
ABGEAM1871AEA 412 CHF
ABGEAM1871A
Anti-PAX6 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PAX6 (paired box 6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR