Anti-HLA DR + DP + DQ Mouse Monoclonal Antibody [clone: CR3/43]

Lieferant: Abcam
AB7856-500
ABCAAB7856-500EA 0 CHF
ABCAAB7856-500
Anti-HLA DR + DP + DQ Mouse Monoclonal Antibody [clone: CR3/43]
Antikörper
Type: Primary
Antigen: HLA DR + DP + DQ
Clonality: monoclonal
Clone: CR3/43
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR