Anti-HLA DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-12]

Lieferant: Abcam
AB28327-100
ABCAAB28327-100EA 0 CHF
ABCAAB28327-100
Anti-HLA DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-12]
Antikörper
Type: Primary
Antigen: HLA DR
Clonality: monoclonal
Clone: MEM-12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR