FMOC-Glu(5/6-TAMRA)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5014
AATB5014EA 769 CHF
AATB5014
FMOC-Glu(5/6-TAMRA)-OH zur biomolekularen Markierung
FMOC-Glu(5/6-TAMRA)-OH
FMOC-Glu(5/6-TAMRA)-OH is a building block for in-sequence peptide Glu labeling by TAMRA.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c021eb733d36b62278628096afdbe100106dbac&vid=40574fc94dd9627d2cb806504e688235f0c496a6&prid=web&clid=SAPDAM