Fluo-5F AM zelldurchlässig

Lieferant: AAT BIOQUEST
Achtung

20560
AATB20560EA 307 CHF
AATB20560
Fluo-5F AM zelldurchlässig
Fluo-5F AM

Fluo-5F is an analog of Fluo-4 with lower calcium-binding affinity (Kd = ~2.3 µM), making it suitable for detecting intracellular calcium levels in the range of 1 µM to 1 mM that would saturate the response of Fluo-4.


  • Improved sensitivity
  • Improved calcium response
  • Cell permeant


Bitte beachten For research use only.

MW: 1100,91 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR