Hoechst 34580 trihydrochloride ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyms: N,N-Dimethyl-4-(6-(4-methylpiperazin-1-yl)-1H,3'H-[2,5'-bibenzo[d]imidazol]-2'-yl)aniline trihydrochloride, Hoechst 34580.3HCl, HOE 34580.3HCl

17537
AATB17537EA 230 CHF
AATB17537
Hoechst 34580 trihydrochloride ≥95%
Hoechst 34580 trihydrochloride

The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy.

Formel: C₂₇H₂₉N₇ . 3HCl
MW: 560,95 g/mol
CAS-Nummer: 911004-45-0
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR