TAQuest™ FAST qPCR Master Mix for TaqMan Probes *High ROX*
Lieferant: AAT BIOQUEST
17292 17293
AATB17292EA 138 CHF
AATB17292 AATB17293
TAQuest™ FAST qPCR Master Mix for TaqMan Probes *High ROX*
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
TAQuest™ FAST qPCR Master mix for TaqMan probes is a ready-to-use 2X solution optimised for qPCR and two-step RT-qPCR well suited to use for TaqMan gene expression assays.
  • Simple and robust protocol
  • The optimised composition ensures PCR specificity and sensitivity with all sample types such as genomic, plasmid, viral and cDNA templates
The master mix is compatible with FAST conditions, thus delivers results within 50 minutes for 40 cycles of PCR in a 20 µl reaction volume.
Bitte beachten Master mix is stable for 12 months when stored frozen (<−15 °C) with minimised light exposure.
Order Now
TECHNISCHE DATEN
Environmentally Preferable
Anwendung qPCR Probe