D(-)-Luciferin Natriumsalz Monohydrate ≥95%

Lieferant: AAT BIOQUEST
Achtung

12509 12510 12511
AATB12509EA 120 CHF
AATB12509 AATB12510 AATB12511
D(-)-Luciferin Natriumsalz Monohydrate ≥95%
D(-)-Luciferin Natriumsalz

Luciferin is the most popular and versatile bioluminescent substrate.

Formel: C₁₁H₇N₂NaO₃S₂·H₂O
MDL Number: MFCD11865368
CAS-Nummer: 103404-75-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9545cea070c2a9fa67002828fa7e826c6e81e881&vid=4acb507e8831cd7b26a5d8d8d9b4ebd3ac5d9a55&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR