Amplite® Colorimetric Asparaginase Activity Assay Kit
Lieferant: AAT BIOQUEST
11800
AATB11800EA 673 CHF
AATB11800
Amplite® Colorimetric Asparaginase Activity Assay Kit
Tests Enzymtests
Designed for consistent and dependable results. The Amplite® colorimetric asparaginase activity assay kit provides a simple and straightforward procedure for measuring asparaginase activity in a variety of biological samples.
  • Robust protocol
  • Absorbance measured at 575 nm
  • Six-month shelf life
One unit (U) is the amount of enzyme that catalyses the reaction of 1 µmol of substrate per minute.
Lieferumfang: Shipped with blue ice.
Bitte beachten Research use only.