5-Carboxyfluorescein 95

Lieferant: AAT BIOQUEST
Achtung

Synonyms: Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-, 5-FAM, 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylic acid, 5-Carboxyfluorescein hydrate

103 105 104
AATB103EA 120 CHF
AATB103 AATB105 AATB104
5-Carboxyfluorescein 95
5-Carboxyfluorescein

5-FAM is the purified single isomer of carboxyfluorescein.

Formel: C₂₁H₁₂O₇.xH₂O
MW: 376.32 g/mol (anhy)
MDL Number: MFCD00036874
CAS-Nummer: 76823-03-5
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cc80d5d97dac535b6103784dd6eed3b35f78805&vid=2db347ed8b66317c345cd473e54fe38375ec62f4&prid=web&clid=SAPDAM

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR