Polyvinylpyrrolidon (K15), BioChemica®

Lieferant: PanReac AppliChem

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

A2258.1000
A2258.1000EA 340 CHF
A2258.1000
Polyvinylpyrrolidon (K15), BioChemica®
Polyvinylpyrrolidon
Formel: (C₆H₉NO)n
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01076626
CAS-Nummer: 9003-39-8

Order Now

Spezifikation Testergebnisse

Assay (from N) 12.0 - 12.8 %
pH (5 %; H2O) 3.0 - 5.0
Heavy metals (as Pb) max. 0.001 %
Water (K.F.) max. 5 %
Solubility (10 %; H2O) clear, colorless to yellowish

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR