1,3-Cyclohexadien 96% stabilisiert

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Cyclohexa-1,3-diene

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A19394.06 A19394.14
A19394.06EA 131 CHF
A19394.06 A19394.14
1,3-Cyclohexadien 96% stabilisiert
1,3-Cyclohexadien

Stabilized with 0.1% BHT


Purity: 96% CAS No.: 592-57-4 Molecular Formula: C6H8 Formula Weight: 80.13 Storage Temperature: 2°C to 8°C Flash Point: -8°(-144°F) Refractive Index: 1.4740 Odour: Unpleasant Melting Point: -89° Boiling Point: 79-81° Beilstein Reference: 506024 UN No.: 3295 MDL No.: MFCD00001532

Formel: C₆H₈
Siedepunkt: 79…81 °C
Melting Pt: –89 °C
Dichte: 0.847
Flash-Pt: –8 °C (–144 °F)
Lager Temperatur: Kühlschrank
MDL Number: MFCD00001532
CAS-Nummer: 592-57-4
EINECS: 209-764-1
UN: 3295
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR