(3,5-Bis(trifluormethyl)phenyl)essigsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

A18922.03 A18922.06
A18922.03EA 45.6 CHF
A18922.03 A18922.06
(3,5-Bis(trifluormethyl)phenyl)essigsäure ≥97%
(3,5-Bis(trifluormethyl)phenyl)essigsäure

Purity: 97% CAS No.: 85068-33-3 Molecular Formula: C10H6F6O2 Formula Weight: 272.15 Melting Point: 121-123° Beilstein Reference: 6813447

Formel: (C₆H₃(CF₃)₂)CH₂COOH
MW: 272.15 g/mol
Siedepunkt: 276
Melting Pt: 121…123 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009908
CAS-Nummer: 85068-33-3
EINECS: 285-295-6

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR