1,5-Diamino-9,10-anthrachinon ≥90%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 1,5-Diaminoanthraquinone

A18840.22 A18840.14
A18840.22EA 408 CHF
A18840.22 A18840.14
1,5-Diamino-9,10-anthrachinon ≥90%
1,5-Diamino-9,10-anthrachinon

Purity: 97% CAS No.: 129-44-2 Molecular Formula: C14H10N2O2 Formula Weight: 238.25 Odour: Odourless Melting Point: ca 320° dec. MDL No.: MFCD00001226

Formel: C₁₄H₁₀N₂O₂
Melting Pt: ∼ 320 °C (dec.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00001226
CAS-Nummer: 129-44-2
EINECS: 204-947-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR